4-Hydroxy-3,5-dimethylbenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A13051.03 A13051.06 A13051.14
A13051.03EA 29.6 EUR
A13051.03 A13051.06 A13051.14
4-Hydroxy-3,5-dimethylbenzoesäure ≥98%
4-Hydroxy-3,5-dimethylbenzoesäure

Purity: 98% CAS No.: 4919-37-3 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 221-226° Beilstein Reference: 2576289

Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Schmelzpunkt: 221…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016536
CAS-Nummer: 4919-37-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR