(4-Chlorphenoxy)essigsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-CPA
A13065.0E
A13065.22
A13065.36
A13065.0EEA
329
EUR
A13065.0E
A13065.22
A13065.36
(4-Chlorphenoxy)essigsäure ≥98%
(4-Chlorphenoxy)essigsäure
Purity: 98+%
CAS No.: 122-88-3
Molecular Formula: C8H7ClO3
Formula Weight: 186.59
Melting Point: 156-161°
Beilstein Reference: 1211804
Formel:
(C₆H₄Cl)OCH₂COOH Molecular Weight: 186,59 g/mol Siedepunkt: 308 °C (1013 hPa) Schmelzpunkt: 156…161 °C Dichte: 1,53 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004305 CAS-Nummer: 122-88-3 EINECS: 204-581-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...