N-(4-Bromphenyl)maleimid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-(4-Bromphenyl)maleinsäureimid
A13119.03
A13119.06
A13119.14
A13119.03EA
26.3
EUR
A13119.03
A13119.06
A13119.14
N-(4-Bromphenyl)maleimid ≥98%
N-(4-Bromphenyl)maleimid
Purity: 98%
CAS No.: 13380-67-1
Molecular Formula: C10H6BrNO2
Formula Weight: 252.07
Melting Point: 127-131°
Beilstein Reference: 513122
Formel:
C₁₀H₆BrNO₂ Molecular Weight: 252,07 g/mol Schmelzpunkt: 127…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00030664 CAS-Nummer: 13380-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...