Maleimid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Maleinsäureimid
A13135.18
A13135.06
A13135.14
A13135.30
A13135.18EA
314
EUR
A13135.18
A13135.06
A13135.14
A13135.30
Maleimid ≥98%
Maleimid
Purity: 98+%
CAS No.: 541-59-3
Molecular Formula: C4H3NO2
Formula Weight: 97.07
Melting Point: 90-94°
Boiling Point: 97-103/5mm
Solubility: Soluble in water
Beilstein Reference: 106910
UN No.: 2923
MDL No.: MFCD00005494
Formel:
C₄H₃NO₂ Molecular Weight: 97.07 g/mol Siedepunkt: 97…103 °C (5 mmHg) Schmelzpunkt: 90…94 °C Dichte: 1.249 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005494 CAS-Nummer: 541-59-3 EINECS: 208-787-4 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...