6-Methyl-5-hepten-2-on ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Methyl-2-hepten-6-on
A13335.14
A13335.22
A13335.36
A13335.14EA
25.8
EUR
A13335.14
A13335.22
A13335.36
6-Methyl-5-hepten-2-on ≥98%
6-Methyl-5-hepten-2-on
Purity: 98%
CAS No.: 110-93-0
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 56°(133°F)
Refractive Index: 1.4390
Melting Point: -67°
Boiling Point: 172-174°
Beilstein Reference: 1741705
UN No.: 1224
MDL No.: MFCD00008905
Formel:
C₈H₁₄O Molecular Weight: 126,2 g/mol Siedepunkt: 172…174 °C Schmelzpunkt: –67 °C Dichte: 0,855 Flammpunkt: 56 °C (133 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008905 CAS-Nummer: 110-93-0 EINECS: 203-816-7 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...