5-Fluoruracil ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,4-Dihydroxy-5-fluorpyrimidin, Fluoruracil, 5-FU
A13456.06
A13456.14
A13456.22
A13456.06EA
50.4
EUR
A13456.06
A13456.14
A13456.22
5-Fluoruracil ≥99%
5-Fluoruracil
Purity: 99%
CAS No.: 51-21-8
Molecular Formula: C4H3FN2O2
Formula Weight: 130.08
Melting Point: ca 282° dec.
Beilstein Reference: 127172
Merck Reference: 14,4181
UN No.: 2811
MDL No.: MFCD00006018
Formel:
C₄H₃FN₂O₂ Molecular Weight: 130.08 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: ∼ 282 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006018 CAS-Nummer: 51-21-8 EINECS: 200-085-6 UN: 2811 ADR: 6.1 III Merck Index: 13,04208 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...