5-Nitro-1H-indazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13478.14
A13478.22
A13478.14EA
28.6
EUR
A13478.14
A13478.22
5-Nitro-1H-indazol ≥98%
5-Nitro-1H-indazole
Purity: 98+%
CAS No.: 5401-94-5
Molecular Formula: C7H5N3O2
Formula Weight: 163.14
Odour: Odourless
Melting Point: 208-211°
Beilstein Reference: 7936
MDL No.: MFCD00005693
Formel:
C₇H₅N₃O₂ Schmelzpunkt: 208…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005693 CAS-Nummer: 5401-94-5 EINECS: 226-451-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...