2-Aminobenzothiazol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Benzothiazol-2-ylamin, Cer(III)-Ionophor II
A13521.22
A13521.36
A13521.22EA
52.4
EUR
A13521.22
A13521.36
2-Aminobenzothiazol 98%
2-Aminobenzothiazole
Purity: 98%
CAS No.: 136-95-8
Molecular Formula: C7H6N2S
Formula Weight: 150.20
Melting Point: 126-129°
Beilstein Reference: 116315
Merck Reference: 14,424
MDL No.: MFCD00005785
Formel:
C₇H₆N₂S Molecular Weight: 150,2 g/mol Siedepunkt: 190…195 °C (0,05 mmHg) Schmelzpunkt: 126…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005785 CAS-Nummer: 136-95-8 EINECS: 205-268-4 Merck Index: 14,424 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...