3-Fluorphthalsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Fluor-1,2-benzoldicarbonsäure
A13656.14
A13656.06
A13656.14EA
286
EUR
A13656.14
A13656.06
3-Fluorphthalsäure ≥97%
3-Fluorphthalsäure
Purity: 97%
CAS No.: 1583-67-1
Molecular Formula: C8H5FO4
Formula Weight: 184.12
Odour: Odourless
Melting Point: ca 170° dec.
Beilstein Reference: 2616684
MDL No.: MFCD00039524
Formel:
C₈H₅FO₄ Molecular Weight: 184.12 g/mol Schmelzpunkt: ∼ 170 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039524 CAS-Nummer: 1583-67-1 EINECS: 216-433-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...