2-Amino-4,6-dimethylpyrimidin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4,6-Dimethylpyrimidin-2-ylamin, 4,6-Dimethylpyrimidin-2-amin
A13674.14
A13674.22
A13674.14EA
28.8
EUR
A13674.14
A13674.22
2-Amino-4,6-dimethylpyrimidin 98%
2-Amino-4,6-dimethylpyrimidine
Purity: 98%
CAS No.: 767-15-7
Molecular Formula: C6H9N3
Formula Weight: 123.16
Melting Point: 150-155°
Beilstein Reference: 110735
Formel:
C₆H₉N₃ Molecular Weight: 123.16 g/mol Schmelzpunkt: 150…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006102 CAS-Nummer: 767-15-7 EINECS: 212-181-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...