5-Isoxazolcarbonsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Isoxazole-5-carboxylic acid

A13739.03 A13739.06
A13739.03EA 66.8 EUR
A13739.03 A13739.06
5-Isoxazolcarbonsäure ≥98%
5-Isoxazolecarboxylic acid

Purity: 98% CAS No.: 21169-71-1 Molecular Formula: C4H3NO3 Formula Weight: 113.07 Melting Point: 141-144° MDL No.: MFCD00156151

Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
Schmelzpunkt: 141…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156151
CAS-Nummer: 21169-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR