2-Isobutylthiazol ≥99%

Lieferant: Thermo Scientific
Achtung

A13759.06 A13759.14
A13759.06EA 62.4 EUR
A13759.06 A13759.14
2-Isobutylthiazol ≥99%
2-Isobutylthiazol

Purity: 99% CAS No.: 18640-74-9 Molecular Formula: C7H11NS Formula Weight: 141.24 Flash Point: 58°(136°F) Refractive Index: 1.4960 Odour: Characteristic Boiling Point: 172-173° Beilstein Reference: 507823 UN No.: 1993 MDL No.: MFCD00005334

Formel: C₇H₁₁NS
Molecular Weight: 141,24 g/mol
Siedepunkt: 172…173 °C
Dichte: 0,998
Flammpunkt: 58 °C (136 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005334
CAS-Nummer: 18640-74-9
EINECS: 242-470-1
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR