4-(Trichlorsilyl)butyronitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Trichlor(3-cyanpropyl)silan, (3-Cyanpropyl)trichlorsilan

A13787.14 A13787.22
A13787.14EA 67.4 EUR
A13787.14 A13787.22
4-(Trichlorsilyl)butyronitril ≥98%
4-(Trichlorsilyl)butyronitril

Purity: 98% CAS No.: 1071-27-8 Molecular Formula: C4H6Cl3NSi Formula Weight: 202.54 Flash Point: 92°(198°F) Refractive Index: 1.4630 Odour: Pungent Boiling Point: 237-238° Beilstein Reference: 906924 UN No.: 2987 MDL No.: MFCD00013832

Formel: NCH₂CCH₂CH₂SiCl₃
Molecular Weight: 202,54 g/mol
Siedepunkt: 237…238 °C
Dichte: 1,28
Flammpunkt: 92 °C (198 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013832
CAS-Nummer: 1071-27-8
EINECS: 213-990-6
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR