3-Chlorbenzoylacetonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-(3-Chlorphenyl)-3-oxopropionsäurenitril, 3-(3-Chlorphenyl)-3-ketopropionsäurenitril

A13913.03 A13913.06
A13913.03EA 26.6 EUR
A13913.03 A13913.06
3-Chlorbenzoylacetonitril ≥98%
3-Chlorbenzoylacetonitril

Purity: 98% CAS No.: 21667-62-9 Molecular Formula: C9H6ClNO Formula Weight: 179.61 Melting Point: 78-83° Beilstein Reference: 2414476

Formel: C₉H₆ClNO
Molecular Weight: 179,61 g/mol
Schmelzpunkt: 78…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067891
CAS-Nummer: 21667-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR