3,5-Dimethylbenzylbromid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: α-Brommesitylen

A14267.06 A14267.14
A14267.06EA 43.8 EUR
A14267.06 A14267.14
3,5-Dimethylbenzylbromid ≥98%
3,5-Dimethylbenzylbromid

Purity: 98% CAS No.: 27129-86-8 Molecular Formula: C9H11Br Formula Weight: 199.10 Melting Point: 36-40° Boiling Point: 109-110°/14mm UN No.: 3261 MDL No.: MFCD00013539

Formel: C₉H₁₁Br
Molecular Weight: 199.09 g/mol
Siedepunkt: 109…110 °C (14 mmHg)
Schmelzpunkt: 36…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013539
CAS-Nummer: 27129-86-8
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR