(Methoxymethyl)triphenylphosphoniumchlorid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A14380.14
A14380.22
A14380.36
A14380.14EA
42.6
EUR
A14380.14
A14380.22
A14380.36
(Methoxymethyl)triphenylphosphoniumchlorid ≥98%
(Methoxymethyl)triphenylphosphonium chloride
Purity: 98+%
CAS No.: 4009-98-7
Molecular Formula: CH3OCH2P(C6H5)3Cl
Formula Weight: 342.81
Storage Temperature: 2°C to 8°C
Melting Point: ca 190° dec.
Beilstein Reference: 924215
MDL No.: MFCD00011800
Formel:
C₂₀H₂₀ClOP Molecular Weight: 342,8 g/mol Schmelzpunkt: ∼ 190 °C (Zersetzung) |
MDL:
MFCD00011800 CAS-Nummer: 4009-98-7 EINECS: 223-664-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...