(4-Methoxyphenoxy)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(4-Methoxyphenoxy)acetic acid

A14419.09 A14419.18
A14419.09EA 42.3 EUR
A14419.09 A14419.18
(4-Methoxyphenoxy)essigsäure ≥98%
(4-Methoxyphenoxy)essigsäure

Purity: 98% CAS No.: 1877-75-4 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 109-113° Beilstein Reference: 1874579

Formel: (C₆H₄(OCH₃))OCH₂COOH
Molecular Weight: 182,18 g/mol
Siedepunkt: 314 °C (1013 hPa)
Schmelzpunkt: 109…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014360
CAS-Nummer: 1877-75-4
EINECS: 217-513-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR