3,5-Dinitro-o-toluylsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Methyl-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-methylbenzoic acid
A14555.14
A14555.22
A14555.36
A14555.14EA
22.9
EUR
A14555.14
A14555.22
A14555.36
3,5-Dinitro-o-toluylsäure ≥98%
3,5-Dinitro-o-toluylsäure
Purity: 98%
CAS No.: 28169-46-2
Molecular Formula: C8H6N2O6
Formula Weight: 226.15
Melting Point: 203-207°
Beilstein Reference: 2622906
Formel:
C₈H₆N₂O₆ Molecular Weight: 226,15 g/mol Schmelzpunkt: 203…207 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007161 CAS-Nummer: 28169-46-2 EINECS: 248-880-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...