Bis(4-methoxyphenyl)methanol ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Bis(4-methoxyphenyl)carbinol, Bis(4-methoxyphenyl)methanol, 4,4'-Dimethoxybenzhydrylalkohol, 4,4'-Dimethoxydiphenylmethanol, 4,4'-Dimethoxy-α-phenylbenzylalkohol
A14604.09
A14604.18
A14604.09EA
30.5
EUR
A14604.09
A14604.18
Bis(4-methoxyphenyl)methanol ≥98%
4,4'-Dimethoxybenzhydrol
Purity: 98+%
CAS No.: 728-87-0
Molecular Formula: C15H16O3
Formula Weight: 244.29
Melting Point: 69-71°
Beilstein Reference: 748532
Formel:
C₁₅H₁₆O₃ Molecular Weight: 244.29 g/mol Siedepunkt: 362 Schmelzpunkt: 69…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008410 CAS-Nummer: 728-87-0 EINECS: 211-975-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...