5-Isoxazolcarbonylchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A14625.03 A14625.06
A14625.03EA 63.7 EUR
A14625.03 A14625.06
5-Isoxazolcarbonylchlorid ≥97%
5-Isoxazolcarbonylchlorid

Purity: 97% CAS No.: 62348-13-4 Molecular Formula: C4H2ClNO2 Formula Weight: 131.52 Refractive Index: 1.4990 Boiling Point: 167-168° UN No.: 3265 MDL No.: MFCD00067868

Formel: C₄H₂ClNO₂
Molecular Weight: 131.52 g/mol
Siedepunkt: 167…168 °C
Dichte: 1.362
Lagertemperatur: Raumtemperatur
MDL: MFCD00067868
CAS-Nummer: 62348-13-4
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR