3-(Methylsulfanyl)propanal ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(Methylthio)propionaldehyd
A14774.09
A14774.18
A14774.30
A14774.09EA
29.7
EUR
A14774.09
A14774.18
A14774.30
3-(Methylsulfanyl)propanal ≥98%
3-(Methylthio)propanal
Purity: 98%
CAS No.: 3268-49-3
Molecular Formula: C4H8OS
Formula Weight: 104.17
Storage Temperature: 2°C to 8°C
Flash Point: 64°(147°F)
Physical Form: Liquid
Density: 1.052
Refractive Index: 1.4825
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Odour: Strong
Melting Point: -58°
Boiling Point: 165-166°
Beilstein Reference: 1739289
UN No.: 2785
MDL No.: MFCD00007022
Formel:
C₄H₈OS Siedepunkt: 165…166 °C Schmelzpunkt: –58 °C Dichte: 1,052 Flammpunkt: 64 °C (147 °F) |
MDL:
MFCD00007022 CAS-Nummer: 3268-49-3 EINECS: 221-882-5 UN: 2785 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...