2-Acetyl-5-bromthiophen ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(5-Brom-2-thienyl)-1-ethanon

A14826.09 A14826.18
A14826.09EA 50.7 EUR
A14826.09 A14826.18
2-Acetyl-5-bromthiophen ≥99%
2-Acetyl-5-bromthiophen

Purity: 99% CAS No.: 5370-25-2 Molecular Formula: C6H5BrOS Formula Weight: 205.07 Melting Point: 93-96° Boiling Point: 103°/4mm MDL No.: MFCD00014528

Formel: C₆H₅BrOS
Molecular Weight: 205.08 g/mol
Siedepunkt: 103 °C (4 mmHg)
Schmelzpunkt: 93…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014528
CAS-Nummer: 5370-25-2
EINECS: 226-363-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR