4-(Benzyloxy)phenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Monobenzon, Hydrochinonmonobenzylether, 4-Hydroxyphenylbenzylether

A14843.0B A14843.14 A14843.22
A14843.0BEA 778 EUR
A14843.0B A14843.14 A14843.22
4-(Benzyloxy)phenol ≥98%
4-Benzyloxyphenol

Purity: 98+% CAS No.: 103-16-2 Molecular Formula: C13H12O2 Formula Weight: 200.24 Melting Point: 118-122° Beilstein Reference: 1958305 Merck Reference: 14,6248 MDL No.: MFCD00002333

Formel: C₁₃H₁₂O₂
Molecular Weight: 200.24 g/mol
Siedepunkt: 328
Schmelzpunkt: 118…122 °C
Dichte: 1.260
Lagertemperatur: Raumtemperatur
MDL: MFCD00002333
CAS-Nummer: 103-16-2
EINECS: 203-083-3
Merck Index: 12,06331

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR