2-Amino-4-methylthiazol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methylthiazol-2-ylamin

A14847.06 A14847.14 A14847.22
A14847.06EA 20.8 EUR
A14847.06 A14847.14 A14847.22
2-Amino-4-methylthiazol ≥98%
2-Amino-4-methylthiazole

Purity: 98+% CAS No.: 1603-91-4 Molecular Formula: C4H6N2S Formula Weight: 114.17 Flash Point: 107°(224°F) Odour: Characteristic Boiling Point: 231-233° Beilstein Reference: 107890 Merck Reference: 14,450 MDL No.: MFCD00005329

Formel: C₄H₆N₂S
Molecular Weight: 114,17 g/mol
Siedepunkt: 231…233 °C
Schmelzpunkt: 42…46 °C
Dichte: 1,202 g/cm³ (50 °C)
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005329
CAS-Nummer: 1603-91-4
EINECS: 216-505-6
Merck Index: 14,450

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR