Methyl-3,5-dihydroxybenzoat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,5-Dihydroxybenzoesäuremethylester
A15224.09
A15224.18
A15224.30
A15224.09EA
43.9
EUR
A15224.09
A15224.18
A15224.30
Methyl-3,5-dihydroxybenzoat ≥98%
Methyl-3,5-dihydroxybenzoat
Purity: 98%
CAS No.: 2150-44-9
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 164-168°
Beilstein Reference: 2091651
Formel:
C₈H₈O₄ Molecular Weight: 168,15 g/mol Siedepunkt: 300 °C (1013 hPa) Schmelzpunkt: 164…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002289 CAS-Nummer: 2150-44-9 EINECS: 218-426-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...