Benzofuran ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzo[b]furan, Cumaron
A15225.09
A15225.18
A15225.30
A15225.09EA
57.8
EUR
A15225.09
A15225.18
A15225.30
Benzofuran ≥99%
Benzofuran
Purity: 99+%
CAS No.: 271-89-6
Molecular Formula: C8H6O
Formula Weight: 118.14
Storage Temperature: 2°C to 8°C
Flash Point: 56°(133°F)
Refractive Index: 1.5660
Odour: Aromatic
Boiling Point: 173-175°
Beilstein Reference: 107704
Merck Reference: 14,1088
UN No.: 1993
MDL No.: MFCD00005847
Formel:
C₈H₆O Molecular Weight: 118,14 g/mol Siedepunkt: 173…175 °C Schmelzpunkt: < –18 °C Dichte: 1,095 Flammpunkt: 56 °C (133 °F) |
MDL:
MFCD00005847 CAS-Nummer: 271-89-6 EINECS: 205-982-6 ADR: 3,III Merck Index: 14,1088 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...