4-Methylthiazol ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
A15428.06
A15428.14
A15428.22
A15428.06EA
36.1
EUR
A15428.06
A15428.14
A15428.22
4-Methylthiazol ≥99%
4-Methylthiazole
Purity: 99%
CAS No.: 693-95-8
Molecular Formula: C4H5NS
Formula Weight: 99.16
Flash Point: 32°(89°F)
Refractive Index: 1.5240
Odour: Characteristic
Boiling Point: 133-134°
Beilstein Reference: 105228
UN No.: 1993
MDL No.: MFCD00005340
Formel:
C₄H₅NS Molecular Weight: 99,16 g/mol Siedepunkt: 133…134 °C Dichte: 1,093 Flammpunkt: 32 °C (89 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005340 CAS-Nummer: 693-95-8 EINECS: 211-764-1 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...