Uracil ≥99%
Lieferant: Thermo Scientific
Synonyme:
2,4-Pyrimidindion, 2,4-Dihydroxypyrimidin, Pyrimidin-2,4(1H,3H)-dion
A15570.0B
A15570.18
A15570.30
A15570.0BEA
575
EUR
A15570.0B
A15570.18
A15570.30
Uracil ≥99%
Uracil
Purity: 99+%
CAS No.: 66-22-8
Molecular Formula: C4H4N2O2
Formula Weight: 112.09
Physical Form: Powder
Appearance: White
Melting Point: ca 330°
Beilstein Reference: 606623
Merck Reference: 14,9850
MDL No.: MFCD00006016
Formel:
C₄H₄N₂O₂ Molecular Weight: 112,09 g/mol Siedepunkt: 379 °C (1013 hPa) Schmelzpunkt: ∼ 330 °C Dichte: 0,99744 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006016 CAS-Nummer: 66-22-8 EINECS: 200-621-9 Merck Index: 14,9850 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...