3-Acetylindol ≥98%
Lieferant: Thermo Scientific
Synonyme:
Indol-3-ylmethylketon, 3-Indolylmethylketon, 3-Indolyl methyl ketone, 1-(3-Indolyl)-1-ethanon
A16221.06
A16221.14
A16221.22
A16221.06EA
37.1
EUR
A16221.06
A16221.14
A16221.22
3-Acetylindol ≥98%
3-Acetylindol
Purity: 98%
CAS No.: 703-80-0
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 186-192°
Beilstein Reference: 122579
Formel:
C₁₀H₉NO Molecular Weight: 159,19 g/mol Siedepunkt: 144…145 °C (10 mmHg) Schmelzpunkt: 186…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005626 CAS-Nummer: 703-80-0 EINECS: 211-875-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...