3-Chlor-4-fluorbenzotrifluorid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Chloro-2-fluoro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluorobenzotrifluoride, 2-Chloro-1-fluoro-4-(trifluoromethyl)benzene

A16670.06 A16670.14
A16670.06EA 69.8 EUR
A16670.06 A16670.14
3-Chlor-4-fluorbenzotrifluorid ≥98%
3-Chlor-4-fluorbenzotrifluorid

Purity: 98% CAS No.: 78068-85-6 Molecular Formula: C7H3ClF4 Formula Weight: 198.55 Flash Point: 42°(108°F) Refractive Index: 1.4355 Boiling Point: 137° Beilstein Reference: 4309386 UN No.: 1993 MDL No.: MFCD00042207

Formel: C₇H₃ClF₄
Molecular Weight: 198,55 g/mol
Siedepunkt: 137 °C
Dichte: 1,490
Flammpunkt: 42 °C (108 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042207
CAS-Nummer: 78068-85-6
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR