(4-tert-Butylphenoxy)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(4-(tert-Butyl)phenoxy)acetic acid

A17017.06 A17017.14 A17017.22
A17017.06EA 17.6 EUR
A17017.06 A17017.14 A17017.22
(4-tert-Butylphenoxy)essigsäure ≥98%
(4-tert-Butylphenoxy)essigsäure

Purity: 98% CAS No.: 1798-04-5 Molecular Formula: C12H16O3 Formula Weight: 208.26 MDL No.: MFCD00021758

Formel: (C₆H₄(C(CH₃)₃))CH₂COOH
Molecular Weight: 208,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021758
CAS-Nummer: 1798-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR