2-Amino-6-brombenzothiazol 98%

Lieferant: Thermo Fisher Scientific
Achtung

A18554.03 A18554.06 A18554.14
A18554.03EA 40.3 EUR
A18554.03 A18554.06 A18554.14
2-Amino-6-brombenzothiazol 98%
2-Amino-6-bromobenzothiazole

Purity: 98% CAS No.: 15864-32-1 Molecular Formula: C7H5BrN2S Formula Weight: 229.10 Odour: Odourless

Formel: C₇H₅BrN₂S
Molecular Weight: 229.1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00152229
CAS-Nummer: 15864-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR