(2Z)-2-Nonen-1-ol 95%
Lieferant: Thermo Scientific
Synonyme:
(Z)-2-Nonen-1-ol
A18842.14
A18842.06
A18842.14EA
163
EUR
A18842.14
A18842.06
(2Z)-2-Nonen-1-ol 95%
cis-2-Nonen-1-ol
Purity: 95%
CAS No.: 41453-56-9
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 101°(214°F)
Refractive Index: 1.4500
Boiling Point: 106°/20mm
MDL No.: MFCD00063210
Formel:
C₉H₁₈O Molecular Weight: 142,24 g/mol Siedepunkt: 106 °C (20 mmHg) Dichte: 0,845 Flammpunkt: 101 °C (214 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063210 CAS-Nummer: 41453-56-9 EINECS: 255-376-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...