1-Chlor-3-fluor-4-iodbenzol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Chlor-1-fluor-2-iodbenzol, 4-Chlor-2-fluoriodbenzol
A19376.06
A19376.14
A19376.06EA
38.8
EUR
A19376.06
A19376.14
1-Chlor-3-fluor-4-iodbenzol ≥98%
1-Chloro-3-fluoro-4-iodobenzene
Purity: 98%
CAS No.: 6797-79-1
Molecular Formula: C6H3ClFI
Formula Weight: 256.45
Flash Point:
Formel:
C₆H₃ClFI Molecular Weight: 256,45 g/mol Siedepunkt: 217 °C Dichte: 1,980 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079729 CAS-Nummer: 6797-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...