2-Fluor-p-anisaldehyd ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Fluor-4-anisaldehyd, 2-Fluor-4-methoxybenzaldehyd
A19726.14
A19726.06
A19726.14EA
225
EUR
A19726.14
A19726.06
2-Fluor-p-anisaldehyd ≥97%
2-Fluor-p-anisaldehyd
Purity: 97%
CAS No.: 331-64-6
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 42-47°
Beilstein Reference: 3237954
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol Schmelzpunkt: 42…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236679 CAS-Nummer: 331-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...