4-Hexinylbenzol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
B20019.03
B20019.06
B20019.03EA
32.3
EUR
B20019.03
B20019.06
4-Hexinylbenzol ≥99%
4-Hexinylbenzol
Purity: 99%
CAS No.: 34298-75-4
Molecular Formula: C6H5(CH2)3C≡CCH3
Formula Weight: 158.25
MDL No.: MFCD00026982
Formel:
(C₆H₅)(CH₂)₃CCCH₃ Molecular Weight: 158,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026982 CAS-Nummer: 34298-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...