Benzylmercaptan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 100-53-8
Molecular Formula: C7H8S
Formula Weight: 124.21
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 1.053
Refractive Index: 1.5760
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Odour: Stench
Melting Point: -30°
Boiling Point: 194-195°
Beilstein Reference: 605864
Merck Reference: 14,9322
UN No.: 2810
MDL No.: MFCD00004867
Formel:
C₇H₈S Siedepunkt: 194…195 °C Schmelzpunkt: –30 °C Dichte: 1,053 Flammpunkt: 70 °C (158 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004867 CAS-Nummer: 100-53-8 EINECS: 202-862-5 UN: 2810 ADR: 6.1,II Merck Index: 14,9322 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...