(2-Methoxyphenoxy)essigsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(2-Methoxyphenoxy)acetic acid
B20672.06
B20672.14
B20672.06EA
16.3
EUR
B20672.06
B20672.14
(2-Methoxyphenoxy)essigsäure ≥98%
(2-Methoxyphenoxy)essigsäure
Purity: 98+%
CAS No.: 1878-85-9
Molecular Formula: C9H10O4
Formula Weight: 182.18
Melting Point: 118-122°
Beilstein Reference: 1874501
Formel:
(C₆H₄OCH₃)OCH₂COOH Molecular Weight: 182.18 g/mol Schmelzpunkt: 118…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014352 CAS-Nummer: 1878-85-9 EINECS: 217-526-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...