2,5-Dimethyl-3-hexin-2,5-diol (±) + meso ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,5-Dihydroxy-2,5-dimethyl-3-hexin

B20889.22 B20889.36
B20889.22EA 32.9 EUR
B20889.22 B20889.36
2,5-Dimethyl-3-hexin-2,5-diol (±) + meso ≥98%
2,5-Dimethyl-3-hexin-2,5-diol

Purity: 98% CAS No.: 142-30-3 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 102°(215°F) Melting Point: 90-94° Boiling Point: 205° Beilstein Reference: 969835 MDL No.: MFCD00004468

Formel: C₈H₁₄O₂
Molecular Weight: 142,2 g/mol
Siedepunkt: 205 °C
Schmelzpunkt: 90…94 °C
Dichte: 0,949
Flammpunkt: 102 °C (215 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004468
CAS-Nummer: 142-30-3
EINECS: 205-533-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR