1,4-Diacetoxybenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: p-Phenylendi(acetat)

B20987.14 B20987.22 B20987.30
B20987.14EA 35 EUR
B20987.14 B20987.22 B20987.30
1,4-Diacetoxybenzol ≥98%
1,4-Diacetoxybenzol

Purity: 98% CAS No.: 1205-91-0 Molecular Formula: C10H10O4 Formula Weight: 194.19 Melting Point: 119-122° Beilstein Reference: 1912233

Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Siedepunkt: 273 °C (1013 hPa)
Schmelzpunkt: 119…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011643
CAS-Nummer: 1205-91-0
EINECS: 214-887-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR