L(+)-Ribose ≥99%

Lieferant: Thermo Scientific

Synonyme: (2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal, L-(+)-Ribose

B21117.03 B21117.14
B21117.03EA 87.8 EUR
B21117.03 B21117.14
L(+)-Ribose ≥99%
L(+)-Ribose

Purity: 99% CAS No.: 24259-59-4 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 85-88° MDL No.: MFCD00167010

Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Siedepunkt: 331 °C (1013 hPa)
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00167010
CAS-Nummer: 24259-59-4
EINECS: 246-110-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR