3-Formylbenzoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Carboxybenzaldehyd, Isophthalaldehydsäure

B21277.03 B21277.06 B21277.14
B21277.03EA 54.1 EUR
B21277.03 B21277.06 B21277.14
3-Formylbenzoesäure ≥97%
3-Formylbenzoesäure

Purity: 97% CAS No.: 619-21-6 Molecular Formula: C8H6O3 Formula Weight: 150.13 Melting Point: 173-175° Beilstein Reference: 2206413

Formel: C₈H₆O₃
Molecular Weight: 150.13 g/mol
Siedepunkt: 306
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039575
CAS-Nummer: 619-21-6
EINECS: 210-585-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR