Dibenzyldiselenid 95%

Lieferant: Thermo Scientific
Gefahr

B21278.03 B21278.06 B21278.14
B21278.03EA 27.9 EUR
B21278.03 B21278.06 B21278.14
Dibenzyldiselenid 95%
Dibenzyldiselenid

Purity: 95% CAS No.: 1482-82-2 Molecular Formula: C6H5CH2Se-SeCH2C6H5 Formula Weight: 340.18 Melting Point: 91-93° Beilstein Reference: 1877745 UN No.: 3283 MDL No.: MFCD00004767

Formel: C₁₄H₁₄Se₂
Molecular Weight: 340,19 g/mol
Schmelzpunkt: 91…93 °C
Dichte: 1,67 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004767
CAS-Nummer: 1482-82-2
UN: 3283
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR