4-Fluorphthalsäureanhydrid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Fluor-1,2-benzoldicarbonsäureanhydrid
B21352.03
B21352.06
B21352.14
B21352.03EA
33.1
EUR
B21352.03
B21352.06
B21352.14
4-Fluorphthalsäureanhydrid ≥98%
4-Fluorphthalsäureanhydrid
Purity: 98%
CAS No.: 319-03-9
Molecular Formula: C8H3FO3
Formula Weight: 166.11
Melting Point: 74-79°
Boiling Point: 147-149°/20mm
MDL No.: MFCD00191363
Formel:
C₈H₃FO₃ Molecular Weight: 166.11 g/mol Siedepunkt: 147…149 °C (20 mmHg) Schmelzpunkt: 74…79 °C Dichte: 1.500 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191363 CAS-Nummer: 319-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...