1,3-Diacetylbenzol ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Benzene-1,3-bis(acetyl)
B21880.03
B21880.06
B21880.03EA
24.7
EUR
B21880.03
B21880.06
1,3-Diacetylbenzol ≥97%
1,3-Diacetylbenzol
Purity: 97%
CAS No.: 6781-42-6
Molecular Formula: C10H10O2
Formula Weight: 162.19
Flash Point:
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162.19 g/mol Siedepunkt: 150…155 °C (15 mmHg) Schmelzpunkt: 35…37 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008740 CAS-Nummer: 6781-42-6 EINECS: 229-842-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...