4-tert-Butylbenzolsulfonylchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-tert-Butylbenzolsulfonsäurechlorid, 4-tert-Butylbenzol-1-sulfonylchlorid, 4-tert-Butylbenzolsulfochlorid

B22174.09 B22174.18
B22174.09EA 46.6 EUR
B22174.09 B22174.18
4-tert-Butylbenzolsulfonylchlorid ≥98%
4-tert-Butylbenzolsulfonylchlorid

Purity: 98% CAS No.: 15084-51-2 Molecular Formula: C10H13ClO2S Formula Weight: 232.73 Odour: Irritating Melting Point: 80-83° Beilstein Reference: 1104346 UN No.: 3261 MDL No.: MFCD00007449

Formel: C₁₀H₁₃ClO₂S
Molecular Weight: 232.73 g/mol
Siedepunkt: 165 °C (18 mmHg)
Schmelzpunkt: 80…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007449
CAS-Nummer: 15084-51-2
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR