1-Brom-3-chlor-4-iodbenzol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Brom-2-chlor-1-iodbenzol

B22185.03 B22185.06 B22185.14
B22185.03EA 48.3 EUR
B22185.03 B22185.06 B22185.14
1-Brom-3-chlor-4-iodbenzol ≥97%
4-Bromo-2-chloro-1-iodobenzene

Purity: 97% CAS No.: 31928-47-9 Molecular Formula: C6H3BrClI Formula Weight: 317.36 Refractive Index: 1.6722 Melting Point: 33-35° Boiling Point: 103-105°/1mm Beilstein Reference: 2435075 MDL No.: MFCD00079709

Formel: C₆H₃BrClI
Molecular Weight: 317,35 g/mol
Siedepunkt: 103…105 °C (1 mmHg)
Schmelzpunkt: 33…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079709
CAS-Nummer: 31928-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR