2-Methyl-6-nitrobenzoxazol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B22415.06 B22415.14
B22415.06EA 78.5 EUR
B22415.06 B22415.14
2-Methyl-6-nitrobenzoxazol ≥98%
2-Methyl-6-nitrobenzoxazole

Purity: 98% CAS No.: 5683-43-2 Molecular Formula: C9H8NO Formula Weight: 146.17 Melting Point: 154-156° MDL No.: MFCD00022848

Formel: C₈H₆N₂O₃
Molecular Weight: 178.15 g/mol
Schmelzpunkt: 154…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022848
CAS-Nummer: 5683-43-2
EINECS: 227-149-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR