2-Amino-5-iodpyrimidin 97%

Lieferant: Thermo Scientific
Achtung

B22705.14 B22705.03 B22705.06
B22705.14EA 308 EUR
B22705.14 B22705.03 B22705.06
2-Amino-5-iodpyrimidin 97%
2-Amino-5-iodopyrimidine

Purity: 97% CAS No.: 1445-39-2 Molecular Formula: C4H4IN3 Formula Weight: 221.00 Melting Point: 217-220° MDL No.: MFCD01075666

Formel: C₄H₄IN₃
Molecular Weight: 221 g/mol
Schmelzpunkt: 217…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075666
CAS-Nummer: 1445-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR