tert-Butylethylether ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Ethoxy-2-methylpropane
B22945.06
B22945.14
B22945.22
B22945.06EA
80.9
EUR
B22945.06
B22945.14
B22945.22
tert-Butylethylether ≥99%
tert-Butylethylether
Purity: 99%
CAS No.: 637-92-3
Molecular Formula: (CH3)3COCH2CH3
Formula Weight: 102.18
Flash Point: -19°(-2°F)
Refractive Index: 1.3750
Melting Point: -94°
Boiling Point: 72-73°
Beilstein Reference: 1731469
Merck Reference: 14,3774
UN No.: 1179
MDL No.: MFCD00009225
Formel:
C₆H₁₄O Siedepunkt: 72…73 °C Schmelzpunkt: –94 °C Dichte: 0,750 Flammpunkt: –19 °C (–2 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009225 CAS-Nummer: 637-92-3 EINECS: 211-309-7 UN: 1179 ADR: 3,II Merck Index: 14,3774 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...